Anti-Respiratory Syncytial Virus Mouse Monoclonal Antibody [clone: 8C5]

NB110-7544
102125-288EA 244.62 USD
102125-288
Anti-Respiratory Syncytial Virus Mouse Monoclonal Antibody [clone: 8C5]
The Respiratory Syncytial Virus Antibody (8C5) from Novus Biologicals is a mouse monoclonal antibody to Respiratory Syncytial Virus. This antibody reacts with virus. The Respiratory Syncytial Virus Antibody (8C5) has been validated for the following applications: ELISA.

Type: Primary
Antigen: Respiratory Syncytial Virus
Clonality: Monoclonal
Clone: 8C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR