Anti-Reg1 Mouse Monoclonal Antibody [clone: 1A4]

Supplier: Novus Biologicals
NBP1-51570
102181-860EA 329 USD
102181-860
Anti-Reg1 Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
The Reg1 Antibody (1A4) from Novus Biologicals is a mouse monoclonal antibody to Reg1. This antibody reacts with human. The Reg1 Antibody (1A4) has been validated for the following applications: ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Reg1
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR