Anti-RDH14 Mouse Monoclonal Antibody [clone: 1A7]

Supplier: Novus Biologicals
NBP2-03667
102234-070EA 663.82 USD
102234-070
Anti-RDH14 Mouse Monoclonal Antibody [clone: 1A7]
Antibodies
The RDH14 Antibody (1A7) from Novus Biologicals is a mouse monoclonal antibody to RDH14. This antibody reacts with human, rat. The RDH14 Antibody (1A7) has been validated for the following applications: Western Blot, Flow Cytometry.

Type: Primary
Antigen: RDH14
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR