Anti-RDH11 Mouse Monoclonal Antibody [clone: 1B4]

Supplier: Novus Biologicals
NBP2-02431
102231-598EA 663.82 USD
102231-598
Anti-RDH11 Mouse Monoclonal Antibody [clone: 1B4]
Antibodies
The RDH11 Antibody (1B4) from Novus Biologicals is a mouse monoclonal antibody to RDH11. This antibody reacts with human. The RDH11 Antibody (1B4) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: RDH11
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR