Anti-Rad51C Mouse Monoclonal Antibody (DyLight 650) [clone: 2H11/6]

Supplier: Novus Biologicals
NB100-177C
102105-926EA 614.15 USD
102105-926
Anti-Rad51C Mouse Monoclonal Antibody (DyLight 650) [clone: 2H11/6]
Antibodies
The Rad51C Antibody (2H11 / 6) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to Rad51C. This antibody reacts with human, mouse, primate, yeast. The Rad51C Antibody (2H11 / 6) [DyLight 650] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Rad51C
Clonality: Monoclonal
Clone: 2H11/6
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Primate, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR