Anti-CD62P Mouse Monoclonal Antibody [clone: HI62P]

NB500-558
102132-674EA 395.38 USD
102132-674
Anti-CD62P Mouse Monoclonal Antibody [clone: HI62P]
The P-Selectin / CD62P Antibody (HI62P) from Novus Biologicals is a mouse monoclonal antibody to P-Selectin / CD62P. This antibody reacts with human. The P-Selectin / CD62P Antibody (HI62P) has been validated for the following applications: Western Blot, Flow Cytometry, CyTOF-ready.

Type: Primary
Antigen: CD62P
Clonality: Monoclonal
Clone: HI62P
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR