Anti-NHE3/SLC9A3 Mouse Monoclonal Antibody [clone: 10A8]

Supplier: Novus Biologicals
NB110-74678 NB110-74678SS
102124-826EA 208 USD
102124-826 102124-814
Anti-NHE3/SLC9A3 Mouse Monoclonal Antibody [clone: 10A8]
Antibodies
The NHE3 / SLC9A3 [p Ser605] Antibody (10A8) from Novus Biologicals is a mouse monoclonal antibody to NHE3 / SLC9A3. This antibody reacts with mouse, rat, opossum, primate, rabbit. The NHE3 / SLC9A3 [p Ser605] Antibody (10A8) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: NHE3/SLC9A3
Clonality: Monoclonal
Clone: 10A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Rat, Opossum, Primate, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR