Anti-NDUFA7 Mouse Monoclonal Antibody [clone: 2G4]

Supplier: Novus Biologicals
NBP2-00564
102227-868EA 663.82 USD
102227-868
Anti-NDUFA7 Mouse Monoclonal Antibody [clone: 2G4]
Antibodies
Mouse Monoclonal NDUFA7 Antibody (2G4). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.

Type: Primary
Antigen: NDUFA7
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR