Anti-NANOG Mouse Monoclonal Antibody [clone: 5A10]

Supplier: Novus Biologicals
NBP1-04320
102156-130EA 564.74 USD
102156-130
Anti-NANOG Mouse Monoclonal Antibody [clone: 5A10]
Antibodies
Mouse Monoclonal Nanog Antibody (5A10). Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:100

Type: Primary
Antigen: NANOG
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR