Anti-NAMPT Mouse Monoclonal Antibody

NBP1-47361
102177-096EA 395.38 USD
102177-096
Anti-NAMPT Mouse Monoclonal Antibody
Mouse Monoclonal NAMPT Antibody (4E11C9). Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.

Type: Primary
Antigen: NAMPT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR