Anti-CDH2 Mouse Monoclonal Antibody [clone: 13A9] (DyLight® 550)

Supplier: Novus Biologicals
NBP1-48309R
102178-468EA 675.69 USD
102178-468
Anti-CDH2 Mouse Monoclonal Antibody [clone: 13A9] (DyLight® 550)
Antibodies
Mouse Monoclonal N Cadherin Antibody (13A9) [DyLight 550]. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 0
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR