Anti-MYL2 Mouse Monoclonal Antibody [clone: 7C9]

Supplier: Novus Biologicals
NBP1-28871
102162-746EA 564.74 USD
102162-746
Anti-MYL2 Mouse Monoclonal Antibody [clone: 7C9]
Antibodies
Mouse Monoclonal Myosin Light Chain 2 Antibody (7C9). Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.

Type: Primary
Antigen: MYL2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR