Anti-MBP Mouse Monoclonal Antibody [clone: BM15]

NB100-73057
102115-892EA 220 USD
102115-892
Anti-MBP Mouse Monoclonal Antibody [clone: BM15]
Mouse Monoclonal Myelin Basic Antibody (BM15). Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.

Type: Primary
Antigen: MBP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR