Anti-Mycobacterium Tuberculosis ESAT6 Mouse Monoclonal Antibody [clone: 11 G4]

Supplier: Novus Biologicals
NBP1-05186
102156-670EA 363 USD
102156-670
Anti-Mycobacterium Tuberculosis ESAT6 Mouse Monoclonal Antibody [clone: 11 G4]
Antibodies
Mouse Monoclonal Mycobacterium Tuberculosis ESAT6 Antibody (11 G4). Tested Applications: Western Blot, ELISA. Tested Reactivity: Bacteria.

Type: Primary
Antigen: Mycobacterium Tuberculosis ESAT6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR