Anti-S100A8 Mouse Monoclonal Antibody [clone: CF-145]

NBP1-02826
102155-562EA 536.92 USD
102155-562
Anti-S100A8 Mouse Monoclonal Antibody [clone: CF-145]
The S100A8 Antibody (CF-145) from Novus Biologicals is a mouse monoclonal antibody to S100A8. This antibody reacts with human. The S100A8 Antibody (CF-145) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 2.5 ?g/mL

Type: Primary
Antigen: S100A8
Clonality: Monoclonal
Clone: CF-145
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR