Anti-PALS1/MPP5 Mouse Monoclonal Antibody [clone: 2D3]

Supplier: Novus Biologicals
NBP2-01880
102230-496EA 663.82 USD
102230-496
Anti-PALS1/MPP5 Mouse Monoclonal Antibody [clone: 2D3]
Antibodies
The PALS1 / MPP5 Antibody (2D3) from Novus Biologicals is a mouse monoclonal antibody to PALS1 / MPP5. This antibody reacts with human, rat, canine. The PALS1 / MPP5 Antibody (2D3) has been validated for the following applications: Western Blot, Flow Cytometry.

Type: Primary
Antigen: PALS1/MPP5
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat, Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR