Anti-MMP11 Mouse Monoclonal Antibody [clone: SL3.05]

Supplier: Novus Biologicals
NBP1-40030
102169-026EA 399 USD
102169-026
Anti-MMP11 Mouse Monoclonal Antibody [clone: SL3.05]
Antibodies
Mouse Monoclonal MMP11 Antibody (LS3.05). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: MMP11
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR