Anti-midA Mouse Monoclonal Antibody [clone: 4A5]

Supplier: Novus Biologicals
NBP2-00820
102228-380EA 663.82 USD
102228-380
Anti-midA Mouse Monoclonal Antibody [clone: 4A5]
Antibodies
Mouse Monoclonal midA Antibody (4A5). Tested Applications: Western Blot, Immunohistochemistry. Tested Reactivity: Human.

Type: Primary
Antigen: midA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR