Anti-MHC1 Mouse Monoclonal Antibody [clone: MEM-E/08]

Supplier: Novus Biologicals
NB500-305
102132-208EA 522.28 USD
102132-208
Anti-MHC1 Mouse Monoclonal Antibody [clone: MEM-E/08]
Antibodies
Mouse Monoclonal MHC Class I Antibody (MEM-E/08). Tested Applications: Flow Cytometry, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: MHC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR