Anti-MGRN1 Mouse Monoclonal Antibody [clone: 3C3]

Supplier: Novus Biologicals
NBP2-00586
102227-912EA 663.82 USD
102227-912
Anti-MGRN1 Mouse Monoclonal Antibody [clone: 3C3]
Antibodies
Mouse Monoclonal MGRN1 Antibody (3C3). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Primate.

Type: Primary
Antigen: MGRN1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR