Anti-MESP1 Mouse Monoclonal Antibody [clone: 1A3]

Supplier: Novus Biologicals
NBP1-51613
102181-944EA 564.74 USD
102181-944
Anti-MESP1 Mouse Monoclonal Antibody [clone: 1A3]
Antibodies
Mouse Monoclonal Mesp1 Antibody (1A3). Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.

Type: Primary
Antigen: MESP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR