Anti-MBD3 Mouse Monoclonal Antibody [clone: 106B691]

Supplier: Novus Biologicals
NB100-56534
102110-080EA 466.46 USD
102110-080
Anti-MBD3 Mouse Monoclonal Antibody [clone: 106B691]
Antibodies
Mouse Monoclonal MBD3 Antibody (106B691). Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MBD3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR