Anti-CD32 Mouse Monoclonal Antibody [clone: AT10]

Supplier: Novus Biologicals
NB100-65338
102114-632EA 564.74 USD
102114-632
Anti-CD32 Mouse Monoclonal Antibody [clone: AT10]
Antibodies
The Fc gamma RII / CD32 Antibody (AT10) from Novus Biologicals is a mouse monoclonal antibody to Fc gamma RII / CD32. This antibody reacts with human, porcine, canine, primate. The Fc gamma RII / CD32 Antibody (AT10) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: CD32
Clonality: Monoclonal
Clone: AT10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Porcine, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR