Anti-CCR9 Mouse Monoclonal Antibody [clone: MM0069-6D10]

Supplier: Novus Biologicals
NBP2-12136
102251-008EA 635.51 USD
102251-008
Anti-CCR9 Mouse Monoclonal Antibody [clone: MM0069-6D10]
Antibodies
The CCR9 Antibody (MM0069-6D10) from Novus Biologicals is a mouse monoclonal antibody to CCR9. This antibody reacts with human. The CCR9 Antibody (MM0069-6D10) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

CyTOF-ready

Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: MM0069-6D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR