Anti-CCR6 Mouse Monoclonal Antibody [clone: 53103.111]

NB120-10398
102126-670EA 352.31 USD
102126-670
Anti-CCR6 Mouse Monoclonal Antibody [clone: 53103.111]
The CCR6 Antibody (53103.111) from Novus Biologicals is a mouse monoclonal antibody to CCR6. This antibody reacts with human. The CCR6 Antibody (53103.111) has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 53103.111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR