Anti-CCR1 Mouse Monoclonal Antibody [clone: MM0061-7B17]

NBP2-12125
102250-986EA 493.85 USD
102250-986
Anti-CCR1 Mouse Monoclonal Antibody [clone: MM0061-7B17]
The CCR1 Antibody (MM0061-7B17) from Novus Biologicals is a mouse monoclonal antibody to CCR1. This antibody reacts with human. The CCR1 Antibody (MM0061-7B17) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CCR1
Clonality: Monoclonal
Clone: MM0061-7B17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR