Anti-CA1 Mouse Monoclonal Antibody [clone: 9D6D7]

Supplier: Novus Biologicals
NBP1-47415
102177-184EA 329 USD
102177-184
Anti-CA1 Mouse Monoclonal Antibody [clone: 9D6D7]
Antibodies
The Carbonic Anhydrase I / CA1 Antibody (9D6D7) from Novus Biologicals is a mouse monoclonal antibody to Carbonic Anhydrase I / CA1. This antibody reacts with human. The Carbonic Anhydrase I / CA1 Antibody (9D6D7) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CA1
Clonality: Monoclonal
Clone: 9D6D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR