Anti-Canine Distemper Virus Mouse Monoclonal Antibody [clone: 7514]

Supplier: Novus Biologicals
NB100-62431
102111-680EA 479.82 USD
102111-680
Anti-Canine Distemper Virus Mouse Monoclonal Antibody [clone: 7514]
Antibodies
The Canine Distemper Virus Antibody (7514) from Novus Biologicals is a mouse monoclonal antibody to Canine Distemper Virus. This antibody reacts with virus. The Canine Distemper Virus Antibody (7514) has been validated for the following applications: ELISA.

Type: Primary
Antigen: Canine Distemper Virus
Clonality: Monoclonal
Clone: 7514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR