Anti-Candida Albicans Mouse Monoclonal Antibody [clone: CDA]

Supplier: Novus Biologicals
NB100-73112
102116-008EA 404.92 USD
102116-008
Anti-Candida Albicans Mouse Monoclonal Antibody [clone: CDA]
Antibodies
The Candida Albicans Antibody (CDA) from Novus Biologicals is a mouse monoclonal antibody to Candida Albicans. This antibody reacts with yeast. The Candida Albicans Antibody (CDA) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Candida Albicans
Clonality: Monoclonal
Clone: CDA
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR