Anti-Calpastatin Mouse Monoclonal Antibody [clone: 1E7]

Supplier: Novus Biologicals
NBP2-01819
102230-374EA 663.82 USD
102230-374
Anti-Calpastatin Mouse Monoclonal Antibody [clone: 1E7]
Antibodies
The Calpastatin Antibody (1E7) from Novus Biologicals is a mouse monoclonal antibody to Calpastatin. This antibody reacts with human. The Calpastatin Antibody (1E7) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Calpastatin
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR