Anti-C20orf30 Mouse Monoclonal Antibody [clone: 2C8]

Supplier: Novus Biologicals
NBP2-03751
102234-238EA 663.82 USD
102234-238
Anti-C20orf30 Mouse Monoclonal Antibody [clone: 2C8]
Antibodies
The C20orf30 Antibody (2C8) from Novus Biologicals is a mouse monoclonal antibody to C20orf30. This antibody reacts with human, monkey. The C20orf30 Antibody (2C8) has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: C20orf30
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR