Anti-BubR1 Mouse Monoclonal Antibody (DyLight 488) [clone: 8G1]

Supplier: Novus Biologicals
NB100-353G
102108-606EA 614.15 USD
102108-606
Anti-BubR1 Mouse Monoclonal Antibody (DyLight 488) [clone: 8G1]
Antibodies
The BubR1 Antibody (8G1) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to BubR1. This antibody reacts with human, mouse. The BubR1 Antibody (8G1) [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: BubR1
Clonality: Monoclonal
Clone: 8G1
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR