Anti-BRCC3 Mouse Monoclonal Antibody [clone: 3B1]

Supplier: Novus Biologicals
NBP1-30429
102163-442EA 564.74 USD
102163-442
Anti-BRCC3 Mouse Monoclonal Antibody [clone: 3B1]
Antibodies
The BRCC3 Antibody (3B1) from Novus Biologicals is a mouse monoclonal antibody to BRCC3. This antibody reacts with human. The BRCC3 Antibody (3B1) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: BRCC3
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR