Anti-BLIMP1/PRDM1 Mouse Monoclonal Antibody [clone: 3H2-E8]

Supplier: Novus Biologicals
NB600-235 NB600-235SS
102133-558EA 577.23 USD
102133-558 102133-570
Anti-BLIMP1/PRDM1 Mouse Monoclonal Antibody [clone: 3H2-E8]
Antibodies
The BLIMP1 / PRDM1 Antibody (3H2-E8) from Novus Biologicals is a mouse monoclonal antibody to BLIMP1 / PRDM1. This antibody reacts with human, mouse, porcine. The BLIMP1 / PRDM1 Antibody (3H2-E8) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: BLIMP1/PRDM1
Clonality: Monoclonal
Clone: 3H2-E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Porcine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR