Anti-Biotin Mouse Monoclonal Antibody [clone: SPM375]

Supplier: Novus Biologicals
NBP1-46960
102176-646EA 244.92 USD
102176-646
Anti-Biotin Mouse Monoclonal Antibody [clone: SPM375]
Antibodies
The Biotin Antibody (SPM375) from Novus Biologicals is a mouse monoclonal antibody to Biotin. This antibody reacts with all species. The Biotin Antibody (SPM375) has been validated for the following applications: Western Blot, Flow Cytometry.

Type: Primary
Antigen: Biotin
Clonality: Monoclonal
Clone: SPM375
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR