Anti-beta 2-Microglobulin Mouse Monoclonal Antibody [clone: B2M-01]

Supplier: Novus Biologicals
NB500-317
102132-232EA 522.28 USD
102132-232
Anti-beta 2-Microglobulin Mouse Monoclonal Antibody [clone: B2M-01]
Antibodies
The beta 2-Microglobulin Antibody (B2M-01) from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, bovine (negative), canine (negative), chicken (negative), mouse (negative), rabbit (negative). The beta 2-Microglobulin Antibody (B2M-01) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Radioimmunoassay.

Radioimmunoassay

Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: B2M-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Bovine (Negative), Canine (Negative), Chicken (Negative), Mouse (Negative), Rabbit (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR