Anti-BAP1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 1G8]

Supplier: Novus Biologicals
NB110-60521H
102124-168EA 614.15 USD
102124-168
Anti-BAP1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 1G8]
Antibodies
The BAP1 Antibody (1G8) [HRP] from Novus Biologicals is a mouse monoclonal antibody to BAP1. This antibody reacts with human. The BAP1 Antibody (1G8) [HRP] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: BAP1
Clonality: Monoclonal
Clone: 1G8
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Within the UCH domain of BAP1.
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR