Anti-AVEN Mouse Monoclonal Antibody [clone: 3G4]

Supplier: Novus Biologicals
NBP1-07098
102157-668EA 564.74 USD
102157-668
Anti-AVEN Mouse Monoclonal Antibody [clone: 3G4]
Antibodies
The AVEN Antibody (3G4) from Novus Biologicals is a mouse monoclonal antibody to AVEN. This antibody reacts with human. The AVEN Antibody (3G4) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50

Type: Primary
Antigen: AVEN
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR