Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]

Supplier: Novus Biologicals
NB100-306 NB100-306SS
102108-468EA 208 USD
102108-468 102108-466
Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]
Antibodies
The ATM [p Ser1981] Antibody (10H11.E12) from Novus Biologicals is a mouse monoclonal antibody to ATM. This antibody reacts with human, mouse, rat, canine (negative). The ATM [p Ser1981] Antibody (10H11.E12) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: ATM
Clonality: Monoclonal
Clone: 10H11.E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Canine (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR