Anti-PNPLA2 Mouse Monoclonal Antibody [clone: 2G2]

NBP1-25852
102160-858EA 395.38 USD
102160-858
Anti-PNPLA2 Mouse Monoclonal Antibody [clone: 2G2]
The PEDF R / PNPLA2 / ATGL Antibody (2G2) from Novus Biologicals is a mouse monoclonal antibody to PEDF R / PNPLA2 / ATGL. This antibody reacts with human, mouse. The PEDF R / PNPLA2 / ATGL Antibody (2G2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:25 - 1:50

Type: Primary
Antigen: PNPLA2
Clonality: Monoclonal
Clone: 2G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR