Anti-ARL11 Mouse Monoclonal Antibody [clone: 1A5]

Supplier: Novus Biologicals
NBP2-01470
102229-676EA 663.82 USD
102229-676
Anti-ARL11 Mouse Monoclonal Antibody [clone: 1A5]
Antibodies
The ARL11 Antibody (1A5) from Novus Biologicals is a mouse monoclonal antibody to ARL11. This antibody reacts with human. The ARL11 Antibody (1A5) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ARL11
Clonality: Monoclonal
Clone: 1A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR