Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Rat IgG Polyclonal Antibody (DyLight® 650)

Supplier: Novus Biologicals
NBP1-76098
102207-174
Goat Anti-Rat IgG Polyclonal Antibody (DyLight® 650)
Goat Polyclonal Goat anti-Rat IgG, H/L Chains Antibody [DyLight 650]. Tested Applications: Flow Cytometry, Immunomicroscopy. Tested Reactivity: Rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype:
Reactivity: Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty