Anti-IgG Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB7149
102135-010EA 225.05 USD
102135-010
Anti-IgG Goat Polyclonal Antibody
Antibodies
Goat Polyclonal Goat anti-Rat IgG, Fc Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry. Tested Reactivity: Rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR