Anti-PLA2G4A Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-52050
102182-586EA 564.74 USD
102182-586
Anti-PLA2G4A Goat Polyclonal Antibody
Antibodies
The PLA2G4A Antibody from Novus Biologicals is a goat polyclonal antibody to PLA2G4A. This antibody reacts with human, mouse. The PLA2G4A Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: PLA2G4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-QEKTFRQQRKEHIR
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR