Anti-CTR9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-68205
102115-510EA 635.51 USD
102115-510
Anti-CTR9 Rabbit Polyclonal Antibody
Antibodies
The CTR9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CTR9. This antibody reacts with human, mouse. The CTR9 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:250

Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR