Anti-CRM1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56493
102110-028EA 279 USD
102110-028
Anti-CRM1 Rabbit Polyclonal Antibody
Antibodies
The CRM1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CRM1. This antibody reacts with human, bovine, canine, primate. The CRM1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml

Type: Primary
Antigen: CRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR