Anti-COX4 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB110-39115B
102121-828EA 614.15 USD
102121-828
Anti-COX4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
The COX4 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to COX4. This antibody reacts with human, mouse, rat, porcine, bovine, drosophila, insect, primate. The COX4 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: COX4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Drosophila, Insect, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR