Anti-Collagen IV alpha 1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB120-6586
102128-176EA 564.74 USD
102128-176
Anti-Collagen IV alpha 1 Rabbit Polyclonal Antibody
Antibodies
The Collagen IV alpha 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Collagen IV alpha 1. This antibody reacts with human, mouse, rat, bovine, mammal. The Collagen IV alpha 1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

There are 2 scientific publications using this antibody.

Type: Primary
Antigen: Collagen IV alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: The development of type-specific antibodies is dependent on NON-DENATURED three-dimensional epitopes.
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Mammal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR