Anti-Clostridium Botulinum Toxin A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-17098
102121-554EA 706.28 USD
102121-554
Anti-Clostridium Botulinum Toxin A Rabbit Polyclonal Antibody
Antibodies
The Clostridium Botulinum Toxin A Antibody from Novus Biologicals is a rabbit polyclonal antibody to Clostridium Botulinum Toxin A. This antibody reacts with bacteria. The Clostridium Botulinum Toxin A Antibody has been validated for the following applications: Western Blot, ELISA, Dot Blot.

Type: Primary
Antigen: Clostridium Botulinum Toxin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR