Anti-CKAP2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-21390
102160-276EA 635.51 USD
102160-276
Anti-CKAP2 Rabbit Polyclonal Antibody
Antibodies
The CKAP2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CKAP2. This antibody reacts with human. The CKAP2 Antibody has been validated for the following applications: Immunoprecipitation, Western Blot (Negative).

Type: Primary
Antigen: CKAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR